Learn More
Medchemexpress LLC Detomidine hydrochloride | 90038-01-0 | 99.8% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB0163A1G
Encompass_Preferred
Detomidine hydrochloride is an imidazole derivative and a potent α2-adrenergic agonist. It induces dose-dependent analgesic effects. Currently, its use is licensed exclusively for horses. The analgesic effects are brought about by the activation of α2 catecholamine receptors, which leads to a negative feedback response that reduces the production of excitatory neurotransmitters. Additionally, Detomidine hydrochloride impacts cardiac and respiratory functions and exhibits an antidiuretic action due to its inhibitory effect on the sympathetic nervous system.
  • Potent α2-adrenergic agonist
  • Induces dose-dependent analgesic effects
  • Licensed for use in horses
  • Inhibits sympathetic nervous system
  • Impacts cardiac and respiratory functions
  • Exhibits antidiuretic action
Catalog No. 50-004-06242