Learn More
Medchemexpress LLC Detomidine hydrochloride | 90038-01-0 | 99.83% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0163A10MM/1ML
Encompass_Preferred
Detomidine hydrochloride is an imidazole derivative and a potent α2-adrenergic agonist. It produces dose-dependent analgesic effects. This product is for research use only and not sold to patients.
  • Potent α2-adrenergic agonist
  • Produces dose-dependent analgesic effects
  • Exhibits cardiac and respiratory effects
  • Has an antidiuretic action
  • Available in solid and solution forms
  • For research use only
Catalog No. 50-004-06085