Learn More
Medchemexpress LLC Detomidine (hydrochloride) | 90038-01-0 | 99.83% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0163A
Encompass_Preferred
Detomidine hydrochloride is an imidazole derivative and a potent α2-adrenergic agonist, demonstrating dose-dependent analgesic effects. It is intended for research use only and its use is currently licensed for horses. This compound produces pain-relieving effects by activating α2 catecholamine receptors, which reduces the production of excitatory neurotransmitters. Additionally, it affects cardiac and respiratory systems and exhibits antidiuretic action due to its inhibition of the sympathetic nervous system.
  • Potent α2-adrenergic agonist
  • Produces dose-dependent analgesic effects
  • Intended for research use only
  • Licensed for use in horses
  • Activates α2 catecholamine receptors
  • Reduces excitatory neurotransmitters
  • Affects cardiac and respiratory systems
  • Exhibits antidiuretic action
Catalog No. 50-004-07406