Learn More
Medchemexpress LLC HY-117287 100mg , Deucravacitinib CAS:1609392-27-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY117287100MG
Encompass_Preferred
HY-117287 100mg Deucravacitinib CAS: 1609392-27-9 Deucravacitinib (BMS-986165) is a highly selective, orally bioavailable allosteric TYK2 inhibitor for the treatment of autoimmune diseases, which selectively binds to TYK2 pseudokinase (JH2) domain (IC50=1.0 nM) and blocks receptor-mediated Tyk2 activation by stabilizing the regulatory JH2 domain. Deucravacitinib inhibits IL-12/23 and type I IFN pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9962