Learn More
Selleck Chemical LLC Deucravacitinib2mg
SDP
Supplier:  Selleck Chemical LLC S88792MG
Encompass_Preferred
Deucravacitinib (BMS-986165) is a highly potent and selective allosteric inhibitor of Tyk2 with a Ki value of 0.02 nM for binding to the Tyk2 pseudokinase domain. It is highly selective against a panel of 265 kinases and pseudokinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3416