Learn More
Azzota Corp Deut Lamp Shimadzu LC-2010
Supplier:  Azzota Corp D2L8169
Encompass_Preferred
New Deuterium Lamp for Shimadzu LC-2010 LC-2010HT LC-2010CHT LC-2010A 2000 hours guaranteed Compatible Manufacture part number 228-37401-91
Catalog No. 50-008-87005