Learn More
Selleck Chemical LLC Deutenzalutamide-E1502-5MG
SDP
Supplier:  Selleck Chemical LLC E15025MG
Encompass_Preferred
Deutenzalutamide (HC-1119 MDV3100) is an androgen receptor (AR) antagonist with an IC50 of 36 nM in LNCaP prostate cells It can be used in a treatment of advanced prostate cancer
Catalog No. 50-313-5771