Learn More
Cambridge Isotope Laboratories Deuterium Oxide "100%" (D, 99.96%), Low Paramagnetic, 7789-20-0, MFCD00044636, 10 g plast
Supplier:  Cambridge Isotope Laboratories DLM117210
Encompass_Preferred
Chemical Formula: D2O, MW: 20.03, 99.5%, Synonyms: Deuterated water; Heavy water (D2O).
Catalog No. NC0354510