Learn More
Wilmad Labglass DEV TANK TLC COMP 75X125MM
Supplier:  Wilmad Labglass LG4595100
Encompass_Preferred
Developing Tank, TLC, Complete. Size: 75mm x 125mm. Heavy wall borosilicate Glass round tank. Can accommodate plate sizes up to 5cm x 10cm. Supplied w/ [ST] 60/21 ground Glass stopper.
Catalog No. 50-806-819