Learn More
Medchemexpress LLC Devimistat-d10 | 2586055-61-8 | C22H18D10O2S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15453S
Encompass_Preferred
Devimistat-d10 is a deuterium-labeled compound, serving as an internal standard and tracer for quantitative analysis in various analytical techniques.
  • Deuterium-labeled compound
  • Suitable as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • High purity (99.79%)
  • Solid, white to off-white appearance
Catalog No. 50-003-75040