Learn More
Cole-Parmer Cole-Parmer Spherical Aluminum and Glass Dewar Flask, 5000 mL
Supplier:  Cole-Parmer 0377430
Encompass
Safely store and transport liquid nitrogen in these fully silvered and evacuated Dewars. Low liquid nitrogen evaporation—only 0.015 liter/hr on 5-liter vessel when covered. Narrow-neck glass vessel sits in a hammer-forged aluminum casing; foam rubber insulation protects against breakage. A heat-insulated cap and carrying handle are included.
Catalog No. NC1456012