Learn More
Biogenex Laboratories Dewax : 1 Liter
SDP
Supplier:  Biogenex Laboratories HK5845KE
Encompass_Preferred
BioGenex deparaffinization solutions are “one-step” products that simultaneously enable the removal of paraffin and allow rehydration of the tissue with a single reagent. In the past, formalin-fixed, paraffin-embedded tissue sections were deparaffinized with highly toxic, noxious chemicals (i.e. xylene, equivalents). BioGenex, a pioneer in the Immunohistochemistry technology, offers xylene-free products that remove the paraffin from mounted tissue slides easily and rapidly.
Catalog No. 50-197-8788
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.