Learn More
Medchemexpress LLC DEX-maleimide | 2754462-35-4 | >99.8% | C29H36FN3O7 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY46225100MG
Encompass_Preferred
DEX-maleimide is a versatile coupling agent designed for the synthesis of conjugates. It plays a crucial role in conjugating single-domain antibodies with various antigens and anti-inflammatory agents, offering significant utility in biochemical research.
  • Can be used to synthesize conjugates
  • Useful in research on autoimmune diseases
  • Facilitates the conjugation of single-domain antibodies with antigens
  • Aids in the conjugation of single-domain antibodies with anti-inflammatory agents
Catalog No. 50-003-58220