Learn More
Medchemexpress LLC Dex-maleimide | 2754462-35-4 | MFCD00064144 | 99.8% | 557.61 g/mol | C29H36FN3O7 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4622510MG
Encompass_Preferred
DEX-maleimide is a maleimide-functionalized dexamethasone derivative used as a coupling reagent for thiol-specific bioconjugation. It is applied to synthesize conjugates such as single-domain antibody (VHH)-antigen or antibody-drug conjugates, and is used in autoimmune disease research. The compound is a solid with high purity and is soluble in DMSO.
Catalog No. 50-002-97885