Learn More
Cayman Chemical Dexamethason-d4 500ug
SDP
Supplier:  Cayman Chemical 22100500 UG
Encompass_Preferred
An internal standard for the quantification of dexamethasone by GC- or LC-MS
Catalog No. 50-298-7288