Learn More
Sigma Aldrich Fine Chemicals Biosciences Dexamethasone, 50-02-2, MFCD00064136, 100mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D1756100MG
Encompass
Molecular Formula: C22H29FO5, Molecular Weight: 392.46, Beilstein Registry Number: 2066651 Assay: ≥98% (HPLC), Solubility: methanol: 25 mg/mL
Catalog No. NC0158347