Learn More
Medchemexpress LLC HY-14648 500mg , Dexamethasone CAS:50-02-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14648500MG
Encompass_Preferred
HY-14648 500mg Dexamethasone CAS: 50-02-2 Dexamethasone (Hexadecadrol) is a glucocorticoid receptor agonist. Dexamethasone also significantly decreases CD11b, CD18, and CD62L expression on neutrophils, and CD11b and CD18 expression on monocytes. Dexamethasone is highly effective in the control of COVID-19 infection. Dexamethasone inhibits production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8986