Learn More
Selleck Chemical LLC Dexamethasone Acetate 10mM/1mL 1177-87-3 NSC 39471
SDP
Supplier:  Selleck Chemical LLC S312410MM/1ML
Encompass_Preferred
Dexamethasone (NSC 39471) is a potent synthetic member of the glucocorticoid class of steroid drugs, and an interleukin receptor modulator that has anti-inflammatory and immunosuppressant effects. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3401