Learn More
U.S. Pharmacopeia Dexamethasone Acetate, 1177-87-3, MFCD00027407, 200mg
Supplier:  U.S. Pharmacopeia 1176506
Encompass
Molecular formula C24H31FO6, Molecular weight 434.51, Melting point/freezing point 460.4 - 464 °F (238 - 240 °C), Synonyms: Dexamethasone 21-acetate
Catalog No. NC1674801