Learn More
Medchemexpress LLC Dexamethasone-d5 | 358731-91-6 | 98.11% | 397.49 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14648S1MG
Encompass_Preferred
Dexamethasone-d5 is a deuterium-labeled form of Dexamethasone. Dexamethasone acts as a glucocorticoid receptor agonist, reducing CD11b, CD18, and CD62L expression on neutrophils and CD11b and CD18 expression on monocytes. It is also effective in controlling COVID-19 infection and inhibiting the production of exosomes containing inflammatory microRNA-155 in lipopolysaccharide-induced macrophage inflammatory responses. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs. This compound is intended for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Appearance: solid
  • Color: white to light yellow
Catalog No. 50-003-63834