Learn More
Medchemexpress LLC Dexketoprofen (trometamol) | 156604-79-4 | >98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0714
Encompass_Preferred
Dexketoprofen (trometamol) is an orally active non-selective COX inhibitor. It is commonly used as an analytical standard for research applications, exhibiting pain-relieving, anti-inflammatory, and anti-cancer effects.
  • Acts as a non-selective COX inhibitor.
  • Provides pain-relieving effects.
  • Demonstrates anti-inflammatory properties.
  • Exhibits anti-cancer activity by inhibiting viability of HeLa, HT-29, and MCF-7 cells.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments like HPLC, GC, and MS.
  • Prevents hyperalgesia and inhibits microglial activation in postoperative pain mouse models.
  • Reduces inflammation and oxidative stress in xylene-induced ear edema rat models.
Catalog No. 50-004-50492