Learn More
Medchemexpress LLC Dexketoprofen (trometamol) | 156604-79-4 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0714100MG
Encompass_Preferred
Dexketoprofen trometamol is an orally active non-selective COX inhibitor. It has pain-relieving, anti-inflammatory, and anti-cancer effects.
  • Pain-relieving effect.
  • Anti-inflammatory effect.
  • Anti-cancer effect.
  • Significantly inhibits the viability of HeLa, HT-29, and MCF-7 cells.
  • Effectively prevents hyperalgesia in a postoperative pain mouse model and inhibits microglial activation.
  • Significantly reduces inflammation and oxidative stress in a xylene-induced ear edema rat model.
Catalog No. 50-004-08183