Learn More
Selleck Chemical LLC Dexketoprofen trometamol E4807-100MG
SDP
Supplier:  Selleck Chemical LLC E4807100MG
Encompass_Preferred
Dexketoprofen trometamol (Dexketoprofen tromethamine salt) is an orally active non-selective inhibitor of COX Dexketoprofen trometamol exhibits analgesic efficacy and can be used in treating acute pain
Catalog No. 50-000-43300