Learn More
Cayman Chemical DexmedetomIdIn 250mg
SDP
Supplier:  Cayman Chemical 39544250 MG
Encompass_Preferred
An α2-AR agonist; selectively binds to α2-ARs over α1-ARs (Kis = 0.015 and 5 nM, respectively); prevents increases in CTnI, as well as reduces myocardial apoptosis, pyroptosis, and ferroptosis in a mouse model of CLP-induced sepsis at 50 UG/kg; induces sedation and decreases mechanical nociception in cats at 40 UG/kg
Catalog No. 50-291-7547