Learn More
Medchemexpress LLC Dexmedetomidine | 113775-47-6 | 99.9% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12719100MG
Encompass_Preferred
Dexmedetomidine is a potent, selective, and orally active agonist of α2-adrenoceptor, with a Ki of 1.08 nM. It demonstrates 1620-fold selectivity against α1-adrenoceptor, exhibiting anxiolysis, sedation, and modest analgesia effects.
  • Potent and selective α2-adrenoceptor agonist (Ki = 1.08 nM)
  • Demonstrates 1620-fold selectivity against α1-adrenoceptor
  • Exhibits anxiolysis, sedation, and modest analgesia effects
  • EC50 of 1.5 nM for agonist activity at human recombinant alpha2A adrenergic receptor
  • Inhibits twitch response in field-stimulated mouse vas deferens with pD2 of 9.0
  • Produces dose-dependent pupillary dilatation in rats
Catalog No. 50-003-40093