Learn More
Medchemexpress LLC Dexrazoxane | 24584-09-6 | 99.88% | 10 MM 1 ML

Supplier: Medchemexpress LLC HYB058110MM/1ML
Dexrazoxane is an intracellular iron chelating agent that reduces the formation of superoxide radicals. It exhibits cardioprotective, anti-inflammatory, antioxidant, anti-tumor, and neuroprotective activities. It inhibits ferroptosis of H9c2 cells by inhibiting HMGB1 and induces DNA damage and apoptosis in human fibrosarcoma cells.
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage for in solvent: -80°C for 6 months, -20°C for 1 month
- Induces DNA breaks, ATF3 accumulation, DNA damage response, and apoptosis in human fibrosarcoma cells
- Inhibits ferroptosis of H9c2 cells by inhibiting HMGB1
- Plays a protective role in rat models of doxorubicin-induced cardiomyopathy
- Provides antioxidant and anti-inflammatory effects in rodent models of Parkinson's disease
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.