Learn More
Medchemexpress LLC Dexrazoxane | 24584-09-6 | MFCD00866449 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0581
Encompass_Preferred
Dexrazoxane is an intracellular iron chelating agent that reduces the formation of superoxide radicals. It exhibits cardioprotective, anti-inflammatory, antioxidant, anti-tumor, and neuroprotective activities, inhibiting ferroptosis and inducing DNA damage and apoptosis in certain cells.
  • Intracellular iron chelating agent
  • Reduces formation of superoxide radicals
  • Exhibits cardioprotective activity
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits anti-tumor activity
  • Exhibits neuroprotective activity
  • Inhibits ferroptosis of H9c2 cells
  • Induces DNA damage and apoptosis in human fibrosarcoma cells
Catalog No. 50-004-50788