Learn More
YSI 3P LifeScience DEXT SUC LACT BUFFR 8/PK MIN 2
Supplier:  YSI 3P LifeScience 098005
Encompass
YSI 2357 Buffer Concentrate Kit for glucose, L-Lactate, dextrose, sucrose, glutamate, glutamine, choline, xylose. 8 envelopes. Reconstitute to make a total of 4 liters. Equipment used with 2900, 2950, 2700, 7100, 2300, 1500. Storage Temperature 0-25C
Catalog No. 50-200-159