Learn More
Immudex Dextramer HLA-A*02:01 Peptide Pool Negative Control, 32 nM, 500 tests, Phycoerythrin (PE)
SDP
Supplier:  Immudex WB05535 PE 500
Encompass
This Immudex Dextramer HLA-A*02:01 Peptide Pool Negative Control is specifically engineered for flow cytometry. It acts as an allele-specific negative control, enabling accurate differentiation between background fluorescence and true antigen-specific T cell staining. The product uses a dextran polymer backbone with a diverse peptide pool and is optimized for sensitive detection in immune monitoring studies.
  • Accurate negative control for flow cytometry.
  • Enhanced specificity with diverse peptide presentation.
  • Compatible with flow cytometry for T cell analysis.
  • Bright phycoerythrin (PE) label for clear signal.
  • Research use only (RUO) for various immunology applications.
Catalog No. NC3667576
Only null left
Add to Cart
Request a Quote