Learn More
Waters Corp Dextran, 40 kDa, Lot Certified, Aqueous Standard
Supplier:  Waters Corp 900114
Encompass_Preferred
This product is a 2 g vial of 40 kDa dextran powder, designed for use as an aqueous standard.
  • Molar mass calibration and A2 validation for DynaPro Plate Reader.
  • Intrinsic viscosity validation for ViscoStar (aqueous mobile phase).
  • Molar mass and A2 validation for Calypso.
  • Compatible with DynaPro Plate Reader III, microViscoStar, ViscoStar II, ViscoStar III, Calypso.
Catalog No. NC2319583