Learn More
LIFESPAN TECHNOLOGIES Dextran Sulfate ELISA Kit, 50 μL sample volume, detection range: LMWDS 0.01 to 3.0 μg/mL, HMWDS 0.003 to 0.3 μg/mL, stable liquid conjugate solution, for buffer and urine samples
Supplier: LIFESPAN TECHNOLOGIES S3900
The Lifespan Technologies S-3900 is an Extended Range Dextran Sulfate ELISA Kit suitable for quantitative detection of low and high molecular weight dextran sulfate. It is ideal for use with buffer and urine samples, and features a stable liquid conjugate solution for user ease. This kit is integral for quality control in various applications, especially in plasma fractioning and vaccine production, where dextran sulfate enhances protein yield in mammalian cell cultures.
- Detection target: Dextran sulfate (LMWDS and HMWDS)
- Sample types: Buffer, urine
- Detection range (HMWDS): 0.003 to 0.3 μg/mL
- Detection range (LMWDS): 0.01 to 3.0 μg/mL
- Conjugate format: Stable liquid solution
- Methodology: ELISA
- Required equipment: Plate reader, absorbance 450 nm
- Sample volume: 50 μL
- Application: In-process quality control in plasma fractioning, virus/vaccine production
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.