Learn More
Medchemexpress LLC Dextran sulfate sodi 500mg | 9011-18-1 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY116282C500MG
Encompass_Preferred
Dextran sulfate sodium salt (MW 35,000-45,000) is a polyanionic sulfated polysaccharide used as a research reagent, commonly to induce experimental colitis in rodents (DSS model). Supplied as a white to off-white solid for laboratory use; powder stability is -20°C (3 years) or 4°C (2 years).
  • Molecular weight 35,000-45,000 Da.
  • Used to induce experimental colitis (DSS model).
  • White to off-white solid appearance.
  • Available in small pack sizes including 500 MG.
  • Store powder at -20°C (3 years) or 4°C (2 years).
  • For research use only; not for human or clinical use.
Catalog No. 50-002-28402