Learn More
Selleck Chemical LLC Dextran sulfate sodium
SDP
Supplier:  Selleck Chemical LLC S69291G
Encompass_Preferred
Dextran sulfate sodium (DSS) is a potent and selective inhibitor of human immunodeficiency virus type 1 (HIV-1) in vitro that inhibits virus adsorption to the host cells Dextran sulfate sodium (DSS) can be used to induce animal models of Enteritis Ulcerative Colitis
Catalog No. 50-000-02158