Learn More
Apexbio Technology LLC Dextran sulfate sodium salt (DSS) 9011-18-1 500g
SDP
Supplier:  Apexbio Technology LLC B8205500000
Encompass_Preferred
Dextran sulfate sodium salt (DSS CAS 9011-18-1) is a water-soluble polyanionic derivative obtained by esterifying dextran with chlorosulfonic acid resulting in approximately 17% sulfur content equivalent to roughly 2 3 sulfate groups per glucose unit DSS induces epithelial damage and intestinal inflammation closely resembling human inflammatory bowel diseases (IBD) including ulcerative colitis Varying its dosage and regimen generates acute chronic or relapsing colitis models for biomedical disease research DSS is also widely utilized in molecular biology procedures such as lipoprotein precipitation enhancing nucleic acid probe hybridization and ribosome isolation
Catalog No. 50-000-63435