Learn More
Sigma Aldrich Fine Chemicals Biosciences Dextran sulfate sodium sal1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences D49111G
Encompass
Dextran sulfate is a sulfated polysaccharide which is negatively charged. It is produced by the esterification of dextran with chlorosulphonic acid. Dextran is used to treat hemorrhage burns surgery and trauma. It improves blood flow and prevents erythrocyte aggregation. Dextran lowers the risk of thrombosis and postoperative pulmonary emboli. It is a ribonuclease inhibitor. Dextran sulfate exhibits antiviral anticoagulant and adjuvant properties.
Catalog No. 50-289-7249