Learn More
Sigma Aldrich Fine Chemicals Biosciences Dextran sulfate sodium sal5G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 428675G
Encompass
Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.
Catalog No. 50-290-1241