Learn More
Alkali Scientific Dextran Sulfate Sodium Salt (DSS), 10 G
Supplier: Alkali Scientific DEX001CP
Dextran Sulfate Sodium Salt (DSS) is a polysaccharide reagent used primarily in molecular biology applications. This 10g bottle is ideal for preparing solutions used in RNA and DNA research, where it is commonly employed in the study of cell interactions and molecular signaling pathways. DSS functions as an anticoagulant and is also used for protein or nucleic acid stabilization in laboratory settings. The reagent is vital for cell culture studies, specifically in understanding the role of heparin-binding molecules in various biological processes. DSS is also utilized for the preparation of immunoassays and assays that examine protein interactions.