Learn More
Alkali Scientific Dextran Sulfate Sodium Salt (DSS), 100 G
SDP
Supplier:  Alkali Scientific DEX002CP
Encompass_Preferred
Dextran Sulfate Sodium Salt (DSS) is a versatile polysaccharide reagent provided in a 100g bottle, widely used in molecular biology for various applications. DSS is crucial in cell culture studies where it acts as a stabilizer for proteins and nucleic acids. It is also a critical component in assays related to molecular signaling, immunology, and drug discovery. DSS enhances the binding of heparin-binding proteins, making it valuable for protein purification processes. Its role in cell interactions and molecular studies aids in understanding complex biochemical processes and assists in the development of bioassays and drug testing systems.
Catalog No. 50-006-61120