Learn More
Alkali Scientific Dextran Sulfate Sodium Salt (DSS), 50 G
Supplier: Alkali Scientific DEX003CP
Dextran Sulfate Sodium Salt (DSS) is a polysaccharide used for molecular biology applications, available in a 50g bottle. DSS is commonly used in cell culture and protein stabilization experiments. The reagent helps prevent oxidative damage and maintains the stability of biological macromolecules. It is also employed in protein purification, RNA or DNA analysis, and as an anticoagulant in various assays. DSS is essential in studies involving heparin-binding proteins and is used to investigate molecular signaling pathways, offering valuable insights in cell biology, immunology, and molecular genetics.