Learn More
Research Products International Corp Dextran Sulfate Sodium Salt, 50 Grams
SDP
Supplier:  Research Products International Corp D2002050.0
Encompass_Preferred
Produced from Dextran 500,000 MW. Tested for use in nucleic acid hybridizations.
Free sulfate content: <0.02%
Catalog No. 50-213-280