Learn More
Medchemexpress LLC Dextran T100 (MW 100,000) | 9004-54-0 | 90000-110000 | 50 G

Supplier: Medchemexpress LLC HY112624O50G
Dextran T100 (MW 100,000) is a polymer of anhydroglucose with an average molecular weight of 100,000. It exhibits good biodegradability and biocompatibility, and is utilized in food, pharmaceutics, cosmetics, and research areas.
- Appearance: Solid
- Color: White to off-white
- Solubility in H2O: ≥ 200 mg/mL
- Store at room temperature for 3 years
- In solvent, store at -80°C for 2 years, -20°C for 1 year
- Ships at room temperature in continental US
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.