Learn More
Medchemexpress LLC Dextran T60 (MW 60,000) | 9004-54-0 | 54000-66000 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY112624D
Encompass_Preferred
Dextran T60 MW 60,000 is a polymer of anhydroglucose with an average molecular weight of 60,000. It demonstrates good biodegradability and biocompatibility, making it suitable for use in food, pharmaceutics, cosmetics, and research applications.
  • Utilized as an extender, anticoagulant, antithrombotic, osmotic agent, and intravenous plasma lubricant in the medical industry.
  • Acts as a cryopreservative for vaccines and organs.
Catalog No. 50-004-39320