Learn More
TARGETMOL CHEMICALS INC DF-1111301 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T271591MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. DF-1111301 is a novel anti-allergic compound with anti-histamine H1 and anti-PAF activity. Purity 99.63%
Catalog No. 50-287-7704