Learn More
Medchemexpress LLC HY-110251 10mg , DFHBI-1T CAS:1539318-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11025110MG
Encompass_Preferred
HY-110251 10mg DFHBI-1T CAS: 1539318-36-9 DFHBI-1T is a membrane-permeable RNA aptamers-activated fluorescence probe (ex/em=472 nm/507 nm). DFHBI-1T binds to RNA aptamers (Spinach, Spinach2, iSpinach, and Broccoli) and causes specific fluorescence and lower background fluorescence. DFHBI-1T is used to image RNA in live cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0520