Learn More
Medchemexpress LLC HY-126073 5mg Medchemexpress, DFP00173 CAS:672286-03-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1260735MG
Encompass_Preferred
Medchemexpress, HY-126073 5mg DFP00173 CAS:672286-03-2 DFP00173 is a potent and selective aquaporin-3 (AQP3) inhibitor. DFP00173 inhibits mouse and human AQP3 with an IC50 of ∼0.1-0.4 μM. DFP00173 is selective for AQP3 over the homologous AQP isoforms AQP7 and AQP9. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9716