Learn More
TARGETMOL CHEMICALS INC DG-041 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1510810MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. DG-041 is an antagonist of high affinity EP3 receptor (IC50s 4.6 nM and 8.1 nM in the binding and FLIPR assay, respectively ). DG-041 inhibits PGE2 facilitation of platelet aggregation and it also crosses the blood-brain barrier. Purity 97.25%
Catalog No. 50-287-8601