Learn More
Cayman Chemical DG-172hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1912810 MG
Encompass_Preferred
An orally available inverse agonist of PPARβ/δ (IC50 = 27 nM), down-regulating transcription of ANGPTL4 in mouse myoblasts (IC50 = 9.5 nM)
Catalog No. 50-298-4869