Learn More
Cayman Chemical dG-PEG 2000 500mg
SDP
Supplier:  Cayman Chemical 37800500 MG
Encompass_Preferred
A PEGylated derivative of DSG; LNPs containing DSG-mPEG(2000) and encapsulating an imidazoquinoline TLR7 and TLR8 agonist and a peptide antigen EAch conjugated to PGA increase the activation of dendritic cells, macrophages, and B cells in the draining lymph node and dendritic cells and B cells in the spleen in mice compared to the non-LNP-encapsulated PGA-conjugated TLR7/8 agonist and peptide antigen.
Catalog No. 50-291-5870