Learn More
Apexbio Technology LLC γDGG 6729-55-1 50mg
SDP
Supplier:  Apexbio Technology LLC B620950
Encompass_Preferred
DGG (CAS 6729-55-1) is a competitive antagonist selective for AMPA-type glutamate receptors where it inhibits excitatory synaptic transmission through direct receptor binding Experimental studies demonstrate that DGG decreases neuronal activity and modulates multiple signaling pathways within the central nervous system Owing to its defined mechanism of action DGG serves as a valuable tool in the investigation of glutamatergic neurotransmission and is broadly utilized in neuroscience research including studies related to central nervous system disorders
Catalog No. 50-000-88106