Learn More
Aviva Systems Biology Corp DGKE Antibody - N-terminal region (ARP45494_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45494_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DGKE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DGKE. Alternate names: DAGK6, DGK, DAGK5ApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-59041